1,3-Dibromo-2,2-dimethoxypropane ≥97.0%

00567-1 00567-2 00567-3
103535-882EA 30.77 USD
103535-882 103535-884 103535-886
1,3-Dibromo-2,2-dimethoxypropane ≥97.0%
1,3-Dibromo-2,2-dimethoxypropane
Formula: BrH₂CC(OCH₃)₂CH₂Br
MW: 261.94 g/mol
MDL Number: MFCD00051792
CAS Number: 22094-18-4

Order Now

Specification Test Results

Purety 97.0%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR