3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid ≥97.0%

Synonyms: 3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid, 3-{[(tert-butoxy)carbonyl]amino}cyclobutane-1-carboxylic acid

00562-1 00562-2 00562-3
103534-434EA 92.31 USD
103534-434 103534-436 103535-874
3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid ≥97.0%
3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid
Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD09864181
CAS Number: 946152-72-3

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR