1,2,3,5-Tetra-O-benzoyl-2-C-methyl-β-D-ribofuranose ≥97.0%

Synonyms: (2S,3R,4R,5R)-2,4-bis(benzoyloxy)-5-[(benzoyloxy)methyl]-3-methyloxolan-3-yl benzoate, (2S,3R,4R,5R)-5-((Benzoyloxy)methyl)-3-methyltetrahydrofuran-2,3,4-triyl tribenzoate

00556-3 00556-1 00556-2
103534-432EA 92.31 USD
103534-432 103535-870 103535-872
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-β-D-ribofuranose ≥97.0%
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose
Formula: C₃₄H₂₈O₉
MW: 580.589 g/mol
MDL Number: MFCD07369658
CAS Number: 15397-15-6

Order Now

Specification Test Results

Purety 97.0%
Appearance

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR