3-Methylpyridazine ≥96.4%

00521-1 00521-2 00521-3
103535-840EA 30.77 USD
103535-840 103535-842 103535-844
3-Methylpyridazine ≥96.4%
3-Methylpyridazine
Formula: C₅H₆N₂
MW: 94.12 g/mol
MDL Number: MFCD00006469
CAS Number: 1632-76-4

Order Now

Specification Test Results

Purety 96.4%
Appearance Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR