1-Methyl-1H-indazole-5-boronic acid ≥97.0%

Synonyms: 1-Methyl-1H-indazol-5-yl-5-boronic acid, (1-methyl-1H-indazol-5-yl)boronic acid

00500-1 00500-2 00500-3
103535-828EA 61.54 USD
103535-828 103535-830 103535-832
1-Methyl-1H-indazole-5-boronic acid ≥97.0%
1-Methyl-1H-indazole-5-boronic acid
Formula: C₈H₉BN₂O₂
MW: 175.9800 g/mol
MDL Number: MFCD09870052
CAS Number: 590418-08-9

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR