tert-Butyl-2,4-dioxopiperidine-1-carboxylate ≥97.0%

Synonyms: tert-Butyl 2,4-dioxopiperidine-1-carboxylate, Piperidine-2,4-dione, N-BOC protected

00493-1 00493-2 00493-3
103535-822EA 30.77 USD
103535-822 103535-824 103535-826
tert-Butyl-2,4-dioxopiperidine-1-carboxylate ≥97.0%
tert-Butyl-2,4-dioxopiperidine-1-carboxylate
Formula: C₁₀H₁₅NO₄
MW: 213.2304 g/mol
MDL Number: MFCD10566071
CAS Number: 845267-78-9

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR