3,3-Difluoropiperidine hydrochloride ≥97.0%

00490-1 00490-2 00490-3
103535-816EA 76.92 USD
103535-816 103535-818 103535-820
3,3-Difluoropiperidine hydrochloride ≥97.0%
3,3-Difluoropiperidine hydrochloride
Formula: C5H10ClF2N
MW: 157.59 g/mol
MDL Number: MFCD03452800
CAS Number: 496807-97-7

Order Now

Specification Test Results

Purety 97.0%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR