(S)-(-)-1-Boc-3-aminopyrrolidine ≥99.0%

Synonyms: (S)-1-Boc-3-Aminopyrrolidine, (S)-(-)-1-BOC-3-aminopyrrolidine, tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate

00471-1 00471-2 00471-3
103535-804EA 30.77 USD
103535-804 103535-806 103535-808
(S)-(-)-1-Boc-3-aminopyrrolidine ≥99.0%
(S)-(-)-1-Boc-3-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
MDL Number: MFCD03419271
CAS Number: 147081-44-5

Order Now

Specification Test Results

Purety 99.0%
Appearance Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR