(R)-3-Hydroxypyrrolidine hydrochloride ≥97.0%

Synonyms: (R)-3-Pyrrolidinol hydrochloride, (3R)-pyrrolidin-3-ol hydrochloride, (R)-3-Hydroxypyrrolidine HCl

00470-1-1 00470-1-2 00470-1-3
103535-798EA 30.77 USD
103535-798 103535-800 103535-802
(R)-3-Hydroxypyrrolidine hydrochloride ≥97.0%
(R)-3-Hydroxypyrrolidine hydrochloride
Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD00191570
CAS Number: 104706-47-0

Order Now

Specification Test Results

Purety 97.0%
Appearance Brown crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR