1-Methylazetidine-3-carboxylic acid ≥97.0%

00463-1 00463-2 00463-3
103535-792EA 92.31 USD
103535-792 103535-794 103535-796
1-Methylazetidine-3-carboxylic acid ≥97.0%
1-Methylazetidine-3-carboxylic acid
Formula: C₅H₉NO₂
MW: 115.1300 g/mol
MDL Number: MFCD11520621
CAS Number: 875629-26-8

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR