(1-Benzhydrylazetidin-3-yl)methanol ≥97.0%

Synonyms: [1-(diphenylmethyl)azetidin-3-yl]methanol

00442-1 00442-2 00442-3
103535-786EA 138.46 USD
103535-786 103535-788 103535-790
(1-Benzhydrylazetidin-3-yl)methanol ≥97.0%
(1-Benzhydrylazetidin-3-yl)methanol
Formula: C₁₇H₁₉NO
MW: 253.35 g/mol
MDL Number: MFCD03407522
CAS Number: 72351-36-1

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR