1-Azetidinecarboxylic acid-3-hydroxyphenylmethyl ester ≥98.0%

Synonyms: Benzyl 3-hydroxyazetidine-1-carboxylate, 1-Cbz-3-Hydroxyazetidine

00429-1 00429-2 00429-3
103535-774EA 69.23 USD
103535-774 103535-776 103535-778
1-Azetidinecarboxylic acid-3-hydroxyphenylmethyl ester ≥98.0%
1-Azetidinecarboxylic acid-3-hydroxyphenylmethyl ester
Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
MDL Number: MFCD09037849
CAS Number: 128117-22-6

Order Now

Specification Test Results

Purety 98.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR