2-(Chloromethyl)pyrimidine hydrochloride ≥96.0%

00381-1-1 00381-1-2 00381-1-3
103535-768EA 69.23 USD
103535-768 103535-770 103535-772
2-(Chloromethyl)pyrimidine hydrochloride ≥96.0%
2-(Chloromethyl)pyrimidine hydrochloride
Formula: C₅H₆Cl₂N₂
MW: 165.02 g/mol
MDL Number: MFCD09834372
CAS Number: 936643-80-0

Order Now

Specification Test Results

Purety 96.0%
Appearance Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR