3-Bromo-1H-pyrazolo[3,4-b]pyridine ≥97.5%

00344-1 00344-2 00344-3
103535-762EA 46.15 USD
103535-762 103535-764 103535-766
3-Bromo-1H-pyrazolo[3,4-b]pyridine ≥97.5%
3-Bromo-1H-pyrazolo[3,4-b]pyridine
Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
MDL Number: MFCD08669476
CAS Number: 68618-36-0

Order Now

Specification Test Results

Purety 97.5%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR