3-(Boc-aminomethyl)azetidine hydrochloride ≥99.7%

Synonyms: tert-butyl N-(azetidin-3-ylmethyl)carbamate

00237-1 00237-2 00237-3
103535-738EA 58.46 USD
103535-738 103535-740 103535-742
3-(Boc-aminomethyl)azetidine hydrochloride ≥99.7%
3-(Boc-aminomethyl)azetidine hydrochloride
Formula: C₉H₁₈N₂O₂
MW: 186.255 g/mol
MDL Number: MFCD01861760
CAS Number: 91188-15-7
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Purety 99.7%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR