3,5-Dimethylisoxazole-4-boronic acid ≥97.1%

Synonyms: (dimethyl-1,2-oxazol-4-yl)boronic acid

00170-1 00170-2 00170-3
103535-714EA 12.31 USD
103535-714 103535-716 103535-718
3,5-Dimethylisoxazole-4-boronic acid ≥97.1%
3,5-Dimethylisoxazole-4-boronic acid
Formula: C₅H₈BNO₃
MW: 140.93 g/mol
MDL Number: MFCD02677945
CAS Number: 16114-47-9

Order Now

Specification Test Results

Purety 97.1%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR