tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate ≥97.0%

Synonyms: Tert-butyl 3-(aminomethyl)azetidine-1-carboxylate, 1-BOC-3-(aminomethyl)azetidine, tert-Butyl 3-(aminomethyl)azetidine-1-carboxylate, 3-Aminomethyl-azetidine-1-carboxylic acid tert-butyl ester

00141-1 00141-2 00141-3
103535-696EA 43.08 USD
103535-696 103535-698 103535-700
tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate ≥97.0%
tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate
Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
MDL Number: MFCD01861762
CAS Number: 325775-44-8

Order Now

Specification Test Results

Purety 97.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR