1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid ≥96.0%

Synonyms: 7-Azaindole-4-carboxylic acid

00136-1 00136-2 00136-3
103535-690EA 100 USD
103535-690 103535-692 103535-694
1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid ≥96.0%
1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.148 g/mol
MDL Number: MFCD08272236
CAS Number: 479553-01-0

Order Now

Specification Test Results

Purety 96.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR