6-Bromo-1H-indazole ≥98.8%

00125-1 00125-2 00125-3
103535-684EA 30.77 USD
103535-684 103535-686 103535-688
6-Bromo-1H-indazole ≥98.8%
6-Bromo-1H-indazole
Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
MDL Number: MFCD03265457
CAS Number: 79762-54-2

Order Now

Specification Test Results

Purety 98.8%
Appearance Brown powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR