4-Bromo-1H-indazole ≥97.0%

Synonyms: 4-Bromoindazole

00105-1 00105-2 00105-3
103535-672EA 53.85 USD
103535-672 103535-674 103535-676
4-Bromo-1H-indazole ≥97.0%
4-Bromo-1H-indazole
Formula: C₇H₅BrN₂
MW: 197.04 g/mol
MDL Number: MFCD05664001
CAS Number: 186407-74-9

Order Now

Specification Test Results

Purety 97.0%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR