7H-Pyrrolo[2,3-d]pyrimidine ≥97.0%

Synonyms: 7-Deazapurine, 7H-Pyrrolo[2,3-d]pyrimidine

00102-1 00102-2 00102-3
103535-666EA 41.54 USD
103535-666 103535-668 103535-670
7H-Pyrrolo[2,3-d]pyrimidine ≥97.0%
7H-Pyrrolo[2,3-d]pyrimidine
Formula: C₆H₅N₃
MW: 119.1240 g/mol
MDL Number: MFCD08234403
CAS Number: 271-70-5

Order Now

Specification Test Results

Purety 97.0%
Appearance Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR