5-Bromo-1H-pyrrolo[2,3-b]pyridine ≥97.0%

Synonyms: 5-Bromo-7-azaindole

00096-1 00096-2 00096-3
103535-660EA 23.08 USD
103535-660 103535-662 103535-664
5-Bromo-1H-pyrrolo[2,3-b]pyridine ≥97.0%
5-Bromo-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₅BrN₂
MW: 197.04 g/mol
MDL Number: MFCD06659677
CAS Number: 183208-35-7

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR