5-Bromo-1H-indazole ≥98.9%

00095-1 00095-2 00095-3
103535-654EA 61.54 USD
103535-654 103535-656 103535-658
5-Bromo-1H-indazole ≥98.9%
5-Bromo-1H-indazole
Formula: C₇H₅BrN₂
MW: 197.03 g/mol
MDL Number: MFCD00839493
CAS Number: 53857-57-1

Order Now

Specification Test Results

Purety 98.9%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR