1-Boc-piperidine-3-carboxylic acid ≥98.0%

Synonyms: N-Boc-DL-nipecotic acid, 1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid, 1-Boc-Nipecotic acid, Boc-Nip-OH, 1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, N-BOC-3-Piperidinecarboxylic acid

00070-1 00070-2 00070-3
103535-648EA 12.31 USD
103535-648 103535-650 103535-652
1-Boc-piperidine-3-carboxylic acid ≥98.0%
1-Boc-piperidine-3-carboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229.3 g/mol
MDL Number: MFCD00673775
CAS Number: 84358-12-3

Order Now

Specification Test Results

Purety 98.0%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR