4-Chloro-1H-pyrazolo[3,4-b]pyridine ≥97.6%

00058-1 00058-2 00058-3
103535-636EA 76.92 USD
103535-636 103535-638 103535-640
4-Chloro-1H-pyrazolo[3,4-b]pyridine ≥97.6%
4-Chloro-1H-pyrazolo[3,4-b]pyridine
Formula: C₆H₄ClN₃
MDL Number: MFCD11846313
CAS Number: 29274-28-0

Order Now

Specification Test Results

Purety 97.6%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR