2-Bromo-5-nitropyridine ≥98.8%

00040-1 00040-2 00040-3
103535-630EA 12.31 USD
103535-630 103535-632 103535-634
2-Bromo-5-nitropyridine ≥98.8%
2-Bromo-5-nitropyridine
Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
MDL Number: MFCD00006222
CAS Number: 4487-59-6

Order Now

Specification Test Results

Purety 98.8%
Appearance Light-Yellow Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR