3,5-Dimethylisoxazole-4-boronic acid pinacol ester ≥99.1%

Synonyms: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 3,5-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole

00029-1 00029-2 00029-3
103535-624EA 30.77 USD
103535-624 103535-626 103535-628
3,5-Dimethylisoxazole-4-boronic acid pinacol ester ≥99.1%
3,5-Dimethylisoxazole-4-boronic acid pinacol ester
Formula: C₁₁H₁₈BNO₃
MW: 223.08 g/mol
MDL Number: MFCD05863910
CAS Number: 832114-00-8

Order Now

Specification Test Results

Purety 99.1%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR