(±)-3-Amino-1-boc-piperidine ≥97.0%

Synonyms: 1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate

00012-1 00012-2 00012-3
103535-612EA 12.31 USD
103535-612 103535-614 103535-616
(±)-3-Amino-1-boc-piperidine ≥97.0%
3-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD01861219
CAS Number: 184637-48-7

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR