Macro Spin Columns, Harvard Apparatus
Supplier: Harvard Apparatus
743901 743904 743907 743900 743902 743903 744302 744309 743906 744303 744308 743905
101167-030PK 1283.08 USD
101167-030 101167-036 101167-042 101167-028 101167-032 101167-034 101167-088 101167-102 101167-040 101167-090 101167-100 101167-038
Macro Spin Columns, Harvard Apparatus
Filters Centrifugal Filters
These macro spin columns are designed for the purification of small samples (10 to 150 µl) in either single column or high-throughput 96-well format with a standard centrifuge.
  • Easy to use
  • High sample recovery and rapid sample preparation time
These spin columns are pre-filled with gel-filtration packing material. The column material binds and purifies the sample according to size and shape, chemical composition, charge or other physiochemical properties.
Applications include DNA purification, protein purification, peptide purification, small molecule and carbohydrate removal , radiolabel removal, nick translation, affinity separation, salt removal, buffer exchange.