Tri-Corner Beakers, United Scientific Supplies
Supplier: United Scientific Supplies
BTC0050 BTC0100 BTC0250 BTC0400 BTC0800 BTC1000
76263-070CS 37.52 USD
76263-070 76263-364 76263-366 76263-368 76263-392 76263-394
Tri-Corner Beakers, United Scientific Supplies
Beakers
Tri-Cornered Beakers are designed to provide three dripless pouring spouts.
  • Graduated in ounces and milliliters
  • Autoclavable
These beakers are graduated in both ounces and milliliters. Available in packs of 100, or as an assortment of 30 beakers featuring 5 of each size.