Adenosine 5′-(α,β-methylene)diphosphate ≥99% (by HPLC), white powder

Synonyms: ADP[α,β-CH2], AMP-CP, α,β-Methyleneadenosine 5′-diphosphate

M3763-25MG M3763-10MG
220197-434EA 572.31 USD
220197-434 220197-433
Adenosine 5′-(α,β-methylene)diphosphate ≥99% (by HPLC), white powder
Adenosine 5′-(α,β-methylene)diphosphate

Adenosine 5′-(α,β-methylene)diphosphate (AMP-CP) is a CD73 inhibitor and is used to study adenosinergic signaling regulation via CD73/ecto-5′-nucleotidase.

Formula: C₁₁H₁₇N₅O₉P₂
MW: 425.23 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00042991
CAS Number: 3768-14-7
EINECS: 223-194-0

Order Now

Specification Test Results

Water (by Karl Fischer) ≤10%
EmM Anhydrous at pH 7.0 14.6 - 15.4
Wave length 259 - 260 nm
Purity (HPLC) ≥99%
Recommended Retest Period 6 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR