Bromotrimethylsilane ≥98% (by titrimetric analysis), clear liquid

Supplier: CHEM IMPEX INTL INC MS
Danger

Synonyms: Trimethylbromosilane, Trimethylsilyl bromide

01939-25G 01939-100G 01939-100G 01939-25G
102616-596EA 36.83 USD
102616-596 102616-598 CH01939-100G CH01939-25G
Bromotrimethylsilane ≥98% (by titrimetric analysis), clear liquid
Bromotrimethylsilane
Formula: C₃H₉BrSi
Density: 1.15…1.16 g/ml
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00000048
CAS Number: 2857-97-8
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Purity (Titration) ≥98%
Infrared spectrum Conforms to reference


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR