tert-Butylchlorodiphenylsilane ≥98% (by GC)

Supplier: CHEM IMPEX INTL INC MS
Danger

Synonyms: tert-Butyldiphenylchlorosilane, tert-Butyl-diphenylchlorosilane

01674-100G 01674-250G 01674-1KG 01674-25G 01674-25G 01674-100G 01674-1KG 01674-250G
102606-568EA 76.55 USD
102606-568 102606-570 102611-090 102616-398 CH01674-25G CH01674-100G CH01674-1KG CH01674-250G
tert-Butylchlorodiphenylsilane ≥98% (by GC)
tert-Butylchlorodiphenylsilane
Formula: C₁₆H₁₉ClSi
Boiling Pt: 90 °C (0.015 mmHg)
Density: 1.057 g/cm³
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000497
CAS Number: 58479-61-1
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Purity (GC) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR