Estradiol, white powder, Cell Culture Grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
beta-Estradiol, 17β-Estradiol
02194565.2
0219456501
0219456505
76204-048EA
72.75
USD
76204-048
76204-050
76204-052
Estradiol, white powder, Cell Culture Grade
Estradiol
β-Estradiol is a potent mammalian estrogenic hormone produced by the ovary. Triggers the production of gonadotropins leading to ovulation.
- Grade : Cell Culture Reagent
- Suitability : Suitable in Cell Culture
- Key Applications : Tumorigenicity Studies
- Product Type : Biochemicals
- Biochemical Category : Lipids
- Chemical Class : Lipids
- xtinction Coefficient : (E1%) EM(280 nm) = 1950 (lit.)
- Optical Rotation : +76.8 (Concentration = 1, Solvent = Dioxane) (Lit.)
- Presentation : White Powder
- Solubility : Almost insoluble in water(50 mg/mL - clear, colorless solution). Freely soluble in alcohol; soluble in acetone, dioxane, other organic solvents; solutions of fixed alkali hydroxides; sparingly soluble in vegetable oils; soluble in methanol:CHCl3 (1:1).
Storage: under nitrogen.
Formula:
C₁₈H₂₄O₂ MW: 272.39 g/mol Melting Pt: 173...179 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003693 CAS Number: 50-28-2 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Melting Point | 173 - 179 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...