Spectinomycin dihydrochloride pentahydrate 500 µg/mg (from Streptomyces species), white powder, Cell Culture Grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate

0219454001 0219454005 0219454025
76203-996EA 60.28 USD
76203-996 76203-998 76204-000
Spectinomycin dihydrochloride pentahydrate 500 µg/mg (from Streptomyces species), white powder, Cell Culture Grade
Spectinomycin dihydrochloride pentahydrate

Spectinomycin is an aminocyclitol antibiotic, closely related to the aminoglycosides, it inhibits protein synthesis (elongation) by interfering with peptidyl tRNA translocation. Mutation in rpsE (the gene for ribosomal protein S5) prevents binding of spectinomycin.


  • Grade : Cell Culture Reagent
  • Specific Activity : 500 µg/mg
  • Suitability : Suitable for cell culture
  • Key Applications : Antibiotic
  • Application Areas : Cell Culture
  • Product Type : Biochemicals
  • Biochemical Category : Antibiotics
  • Melting Point : 205-207 °C(Lit.)
  • Presentation : White Powder
  • Loss on Drying : <20%
  • Solubility : Soluble in water (50 mg/mL), methanol, propyleneglycol; very soluble benzene, chloroform, ethanol and acetone.

Formula: C₁₄H₂₄N₂O₇·2HCl·5H₂O
MW: 495.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00150886
CAS Number: 22189-32-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c37f7aceb0cdcac7201befde420d656972a80e8c&vid=276f89cf97ceafe394335cb7dc02d57cb2fedaa6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Source Streptomyces species
Activity 500 µg/mg
FTIR Conforms to Standard
Loss on Drying <20%
Optical Rotation Positive

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR