Penicillin G potassium salt, white powder, Cell Culture Grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 6-(2-Phenylacetamido)penicillanic acid potassium salt, Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin G potassium, Potassium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Benzylpenicillin potassium salt

0219453601 0219453610 0219453625 0219453680
76204-030EA 126.72 USD
76204-030 76204-032 76204-034 76204-036
Penicillin G potassium salt, white powder, Cell Culture Grade
Penicillin G potassium salt

Penicillin G potassium Salt is a salt of Penicillin G. Penicillin G is a hydrophobic, β-lactam antibiotic produced by Penicillium spp. Penicillin G is also known as benzylpenicillin and contains a 6’ phenylacetyl side chain. In acid aqueous solutions, Penicillin is reported to decompose into penillic acid and penilloic acid.


  • Grade : Cell culture reagent
  • Potency : 1,500 - 1,700 u/mg
  • Suitability : Suitable for cell culture.
  • Key Applications : Cell culture reagent; Antibiotic; Inhibitor of bacterial cell wall synthesis
  • Product Type : Biochemicals
  • Biochemical Category : Antibiotics
  • Chemical Class : Antibiotics
  • Presentation : White Powder
  • pH 5 : - 7.5 (3% aq soln)
  • Solubility : Soluble in water(100 mg/mL).

Formula: C₁₆H₁₇KN₂O₄S
MW: 372.49 g/mol
Melting Pt: 214...217 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00036193
CAS Number: 113-98-4
Merck Index: 12,07225

Order Now

Specification Test Results

Identity Test Passes
pH (3% aq soln) 5 - 7.5
Potency 1,500 - 1,700 u/mg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR