Paromomycin sulfate ≥97%, white powder, Cell Culture Grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0219453501
0219453505
76203-982EA
59.68
USD
76203-982
76203-984
Paromomycin sulfate ≥97%, white powder, Cell Culture Grade
Paromomycin sulfate
Paromomycin Sulfate is an aminoglycoside antibiotic which inhibits protein initiation.
- Grade : Cell culture reagent
- Purity : ≥97%
- Potency : >675 µg/mg
- Key Applications : Antibiotic
- Application Areas : Molecular Biology
- Product Type : Biochemicals
- Chemical Class : Antibiotics
- Presentation : White Powder
- Format : Powder
- Solubility : Soluble in water (25% w/v - light opalescent solution).
Formula:
C₂₃H₄₅N₅O₁₄·H₂SO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00079278 CAS Number: 1263-89-4 |
|
Specification Test Results
Identity Test | Passes |
Purity | >97% |
FTIR | Conforms to Reference |
Potency | >675 mg/mg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...