Pyrimethamine
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
5-(4-Chlorophenyl)-6-ethylpyrimidine-2,4-diamine, 5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine
0219418001
0219418005
0219418025
76204-338EA
72.65
USD
76204-338
76204-340
76204-342
Pyrimethamine
Pyrimethamine
Pyrimethamine is a diamino-pyrimidine and folic acid antagonist.
- Key Applications : Antiprotozoal compound
- Application Areas : Antibiotic
- Product Type : Biochemicals
- Presentation : White powder
- Solubility : Slightly soluble in ethanol (room temperature: 9 mg/mL - clear, colorless to faint yellow solution; boiling ethanol: 25 mg/mL) or in dilute Hydrochloric acid (5 mg/mL).
Formula:
C₁₂H₁₃ClN₄ MW: 248.71 g/mol Melting Pt: 240...242 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00057350 CAS Number: 58-14-0 |
Specification Test Results
Identity Test | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...