Hydrochloric acid ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0219405480 0219405490 0219405491
76204-098EA 59.32 USD
76204-098 76204-100 76204-102
Hydrochloric acid ACS
Hydrochloric acid

  • Grade ACS Reagent Grade
  • Assay 36.5 - 38%
  • Product Type Inorganics
  • Biochemical Category Acids
  • Vapor Density 1.267 (air = 1)(Lit.)
  • Vapor Pressure 3.23 psi ( 21.1 °C) (Lit.); 7.93 psi ( 37.7 °C) (Lit.)
  • Presentation Clear, Colorless Liquid
  • Color (APHA) ≤10
  • Heavy Metals Lead ≤1 ppm
  • Residue on ignition ≤5 ppm
  • NOTES Bromide (Br) ≤0.005%
  • Sulfate (SO4) ≤1 ppm
  • Sulfite (SO3) ≤1 ppm
  • Free Chlorine (Cl) ≤1 ppm
  • Ammonium (NH4) ≤3 ppm
  • Arsenic (As) ≤0.01 ppm
  • Iron (Fe) ≤0.2 ppm
  • Extractable Organic Substances: ≤5 ppm

Formula: HCl
Boiling Pt: >100 °C (lit.)
Melting Pt: –114.4 °C (lit.)
Density: 1.161...1.19 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: ,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d242f6967f29026d2f2b6323fe0f29e1e9e7a8d1&vid=93b3fafdf986e766c3a56e3ac9d0ddcf49c169cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 36.5 - 38%
Appearance Free from suspended
matter or sediment,
Color (APHA) <10
Residue after Ignition <5 ppm
Bromide (Br) <0.005%
Sulfate (SO4) <1 ppm
Free Chlorine (CI) <1 ppm
Ammonium (NH4) <3 ppm
Arsenic (As) <0.01 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR