Hexamethylenetetramine, white crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Hexamethylenetetramine, Hexamine, Urotropine, Formin

0219405083 0219405091
76204-094EA 58.98 USD
76204-094 76204-096
Hexamethylenetetramine, white crystalline powder ACS
Hexamethylenetetramine

Hexamethylenetetramine is a tetraamine compound with a structure analogous to adamantane.


Hexamethylenetetramine is used for amino acid preparation. It is also used in synthesis as a formylation agent and is employed in preparing specialised materials. It can be used as raw material in the synthesis of blue emitting boron carbon oxynitride (BCNO) phosphors and zinc oxide (ZnO) nanorods. Catalyst in the synthesis 2-amino-4H-pyran derivatives. Surfactant in the synthesis of HMTA capped transition metals doped ZnS nanoparticles. Precipitant and template in the synthesis of Ni(OH)₂.


Solubility : Soluble in cold water, chloroform, alcohol; Insoluble in diethyl ether.


Precaución: Research Use Only (RUO).

Formula: C₆H₁₂N₄
MW: 140.19 g/mol
Melting Pt: 280 °C (subl., lit.)
Density: 1.33 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00006895
CAS Number: 100-97-0
UN: 1328
ADR: ,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaec0198460b370c901bacd6f62ec0558c921a37&vid=eaec0198460b370c901bacd6f62ec0558c921a37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (dried basis) >99%
Loss on Drying <2.0%
Residue after Ignition <0.1%
Heavy Metals (as Pb) <0.001%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR