Glycine, white crystalline powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
0219404983
0219404991
0219404992
0219404950
76204-088EA
152.24
USD
76204-088
76204-090
76204-092
76204-322
Glycine, white crystalline powder ACS
Glycine
Glycine is a non-essential amino acid. Only amino acid with no asymmetric carbon. It is a major inhibitory neurotransmitter.
- Grade : ACS Reagent Grade
- Assay : ≥98.5%
- Key Applications : PAGE compound
- Application Areas : Protein Electrophoresis & Blotting
- Product Type : Biochemicals
- Biochemical Category : Amino Acids & Peptides
- Chemical Class : Amino Acids & Peptides
- Format : Crystalline Powder
- pKa :2.35 at 25 °C (Lit.)
- Heavy Metals : Lead (Pb): ≤0.002%
- Residue on ignition: ≥0.1%
- NOTES Chloride (Cl) : ≤0.005%, Sulfate (SO4): ≤0.005%, Ammonium (NH4): ≤0.005%
- Solubility : Soluble in water (250 mg/mL at 25 °C; 672 mg/mL at 100 °C); slightly soluble in alcohol and ethanol(95%); almost insoluble in ether.
Formula:
NH₂CH₂COOH MW: 75.07 g/mol Melting Pt: 240 °C (dec., lit.) Density: 1.16 g/mL (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 |
|
Specification Test Results
Assay | > 98.5% |
Residue after ignition | Not More Than 0.1% |
Heavy Metals (as Pb) | Not More Than 0.002% |
Chloride (Cl) | Not More Than 0.005% |
Sulfate (SO4) | Not More Than 0.005% |
Substances darkened by sulfuric acid |
Passes Test |
Hydrolyzable substances | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...