Lithium chloride ≥99%, white crystalline powder for molecular biology
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Lithium Chloride is a metal halide utilized in a variety of assays to study cell-fate, neurobiology and antiviral properties.
Lithium Chloride is used as an electrolyte for low temperature dry battery cells and as an oxidation catalyst. It is a solubilizer for polyamides and cellulose when used with amide solvents, and is a chlorinating agent for steroid substrates. It has also been utilized in: Large scale plasmid DNA isolation without ultracentrifugation; Protein extraction and protein crystallization; Crystallization of other biological structures, including vitamin B12-RNA aptamer and the L-A virus particle; Inhibits the expression and secretion of insulin-like growth factor-binding protein-1 in H4-II-E cells; Used in the synthesis of beta-substituted alpha-amino acid derivatives; May be used to selectively precipitate RNA.
Formula:
LiCl MW: 42.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011078 CAS Number: 7447-41-8 |
Specification Test Results
Identity Test | Passes |
Purity | >99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...