N-Lauroylsarcosine sodium salt ≥97%, white powder, Ultrapure

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate

0219400990 0219400950 0219400980
76203-950EA 561.07 USD
76203-950 76204-308 76204-310
N-Lauroylsarcosine sodium salt ≥97%, white powder, Ultrapure
N-Lauroylsarcosine sodium salt

N-Lauroylsarcosine is a derivative of sarcosine . Its sodium salt is anionic surfactant.


  • Grade : Ultra pure
  • Purity : ≥97%
  • Key Applications : Anionic detergent; Inhibitor of Bacterial flora of human saliva/gut
  • Product Type : Biochemicals
  • Biochemical Category : Surfactants
  • Chemical Class : Surfactants
  • Extinction Coefficient : (E1%) EM = 3 (280 nm)(Lit.)
  • UV/Visible Absorbance : λ max:220 nm, 265 nm(Lit.)
  • Presentation : White Powder
  • pH : 7.5 - 8.5(10% aq soln)
  • Aggregation Number : 2(Lit.)
  • Critical Micelle Concentration (CMC) : 14.57 mM at 30 °C(Lit.)
  • Solubility : Soluble in water (100 mg/mL - clear to slightly hazy, colorless solution).

Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MW: 293.38 g/mol
Flash Pt: 267 °C (512.6 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
UN: 2811
ADR: ,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b7815e4a8efd2ddde5d98aa7b518ea0d009ec4f&vid=b33c5ca726cf4eb3d69fc16f8dd1fe434e20fbb0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity >97%
pH (10% aq soln) 7.5 - 8.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR