Ammonium acetate ≥98%, powder for molecular biology
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Acetic acid ammonium salt
0219400080
0219400090
76204-066EA
44.23
USD
76204-066
76204-068
Ammonium acetate ≥98%, powder for molecular biology
Ammonium acetate
Ammonium, Biochemicals, Biological Buffers, Buffer / Buffer Salts, Buffers / Buffer Salt, Chemical Synthesis, Inorganic Salts, Life Science Reagents for Cloning, Life Science Reagents for PCR, Life Science Reagents for Protein Expression and Purification, Molecular Biology, Molecular Biology Reagents, Research Essentials, Synthetic Reagents
- Grade : Molecular Biology Reagent
- Purity : ~98%
- Key Applications : Purification and precipitation of DNA; protein crystallization; HPLC and MS analysis
- Application Areas : Purification and precipitation of DNA, protein crystallization, HPLC and MS analysis
- Product Type : Inorganics
- Format : Powder
- pH : (5% aq soln) 6 - 8
- Solubility : Soluble in water (4 g/4 mL-clear, colorless solution).
Storage: under nitrogen.
Formula:
H₃CCOONH₄ MW: 77.08 g/mol Boiling Pt: (dec., lit.) Melting Pt: 114 °C (237 °F) (lit.) Density: 1.17 (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00013066 CAS Number: 631-61-8 UN: 3077 ADR: 9,III |
Specification Test Results
Identity Test | Passes |
Purity | ~98% |
pH (5% aq soln) | 6 - 8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...