Zeolite, brown pellets

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0219390280 0219390290
76204-024EA 93.82 USD
76204-024 76204-026
Zeolite, brown pellets
Zeolite

  • Key Applications : Molecular Seive
  • Product Type : Biochemicals
  • Presentation : Brown Pellets

Formula: Na₂O·Al₂O₃·xSiO₂·yH₂O
Storage Temperature: Ambient
MDL Number: MFCD00132601
CAS Number: 1318-02-1

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR