Calcium acetate monohydrate, white powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Calcium di(acetate) monohydrate, Calcium acetate hydrate, Acetic acid calcium salt monohydrate
0219382190
0219382180
76204-022EA
217.04
USD
76204-022
76204-132
Calcium acetate monohydrate, white powder ACS
Calcium acetate monohydrate
White Powder
- Grade : ACS
- Product Type : Inorganics
- Chemical Class : Salts
- Presentation : White Powder
Formula:
(H₃CCOO)₂Ca·1H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150019 CAS Number: 5743-26-0 |
Specification Test Results
Identity Test | Passes |
Purity | >99.0% |
FTIR | Conforms to Standard |
pH (0.2 M aq soln) | 7.4 - 7.6 |
Heavy Metals | <0.005% |
Alkalinity | Passes |
Chloride | <0.001% |
Barium | <0.01% |
Iron | <0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...