Calcium phosphate tribasic, orange powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0219380525 0219380580 0219380590
76204-126EA 65.81 USD
76204-126 76204-128 76204-130
Calcium phosphate tribasic, orange powder
Calcium phosphate tribasic

  • Key Applications : Used in manufacture of fertilizers, polishing and dental powders, porcelain and pottery, and enameling; Utilized to engineer new biomaterials for applications : such as bone grafts and fillers.
  • Application Areas : Industrial
  • Product Type : Inorganics
  • Chemical Class : Salts
  • Presentation : Orange Powder
  • Solubility : Soluble in 1 M Hydrochloric acid (50 mg/mL - clear to slightly hazy, colorless to slight yellowish-green solution).

Formula: Ca₅HO₁₃P₃
Melting Pt: 1100 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00010904
CAS Number: 12167-74-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de581a0357e9273c0524dd76dcab4ee749d61b76&vid=de581a0357e9273c0524dd76dcab4ee749d61b76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR