Calcium bis(dihydrogenorthophosphate), white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219380350 0219380380 0219380390
IC0219380350EA 63.25 USD
IC0219380350 IC0219380380 IC0219380390
Calcium bis(dihydrogenorthophosphate), white powder
Calcium bis(dihydrogenorthophosphate)

  • Key Applications : Mineral supplement; Fertilizer; Leavening agent
  • Application Areas : Molecular Biology
  • Product Type : Inorganics
  • Biochemical Category : Biochemicals
  • Chemical Class : Salts
  • Vapor Pressure : Zero
  • Presentation : White Powder
  • Format : Powder
  • Solubility : Soluble in 1 M Hydrochloric acid (50 mg/mL - clear, colorless solution), dilute HNO3, acetic acid; moderately soluble in water.

Formula: H₄CaO₈P₂
MW: 234.05 g/mol
Melting Pt: 203 °C (dec., lit.)
Density: 2,22 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00010898
CAS Number: 7758-23-8

Order Now

Specification Test Results

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR