CleanMax® Clean Manufactured Disposable Frocks/Lab Coats, Lakeland Industries
Supplier: Lakeland
CTL191CM-MD CTL191CM-LG CTL191CM-XL CTL191CM-2X CTL191CM-3X CTL191CM-4X CTL191CM-5X
76193-202CS 672.22 USD
76193-202 76193-204 76193-206 76193-208 76193-210 76193-318 76193-320
CleanMax® Clean Manufactured Disposable Frocks/Lab Coats, Lakeland Industries
Disposable Garments
Clean manufactured disposable frocks and lab coats are individually bagged and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • Latex-free
  • Mandarin collar
  • Tunneled elastic wrists with thumb loops
  • Zipper closure
  • No pockets
  • Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunneled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certificaciones: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.