Sodium carbonate, anhydrous 99.5-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., NF, FCC, E500, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
13418-5KG
13418-1KG
BJ13418-5KGEA
291.9
USD
BJ13418-5KG
BJ-13418-1KG
BJ-13418-2.5KG
Sodium carbonate, anhydrous 99.5-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., NF, FCC, E500, Fluka™
Sodium carbonate
Formula:
Na₂CO₃ Melting Pt: 854 °C Density: 2.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 |
|
Specification Test Results
Assay (calculated to the dried substance) | 99.5 - 100.5 % |
Loss on drying (300°C) | Max. 0.5 % |
Loss on drying (105°C, 4 h) | Max. 0.5 % |
Arsenic (As) | Max. 2 ppm |
Copper (Cu) | Max. 0.002 % |
Iron (Fe) | Max. 10 ppm |
Mercury (Hg) | Max. 1 ppm |
Lead (Pb) | Max. 2 ppm |
Zinc (Zn) | Max. 0.002 % |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 100 ppm |
Sulfate (SO₄) | Max. 50 ppm |
Alkali hydroxide, -bicarbonates | complying |
Appearance of the solution | complying |
Residual solvents | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...